Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYATL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696397.3 | XP_005696454.2 | 2 - 3 | 207 | 95 | |
GenBank mRNA | JO397937.2 | - | 2 - 3 | 213 | 95 |
Gene Symbol: LOC108635060 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045363.1 | XP_017900852.1 | 2 - 3 | 194 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |