Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052929.1 | XP_017908418.1 | 2 - 3 | 310 | 71 | |
XM_018052930.1 | XP_017908419.1 | 2 - 3 | 284 | 71 | ||
GenBank mRNA | JT704089.1 | - | 3 - 4 | 573 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |