Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047761.1 | XP_017903250.1 | 4 - 5 | 1586 | 93 | |
XM_018047762.1 | XP_017903251.1 | 5 - 6 | 1214 | 93 | ||
XM_018047763.1 | XP_017903252.1 | 4 - 5 | 1041 | 93 | ||
GenBank mRNA | JT666426.1 | - | 5 - 6 | 908 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |