Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054445.1 | XP_017909934.1 | 4 - 5 | 2152 | 63 | |
XM_018054446.1 | XP_017909935.1 | 4 - 5 | 2149 | 63 | ||
XM_018054447.1 | XP_017909936.1 | 4 - 5 | 2152 | 63 | ||
GenBank mRNA | JO558412.2 | - | 2 - 3 | 207 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |