Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL20RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053252.1 | XP_017908741.1 | 3 - 4 | 577 | 105 | |
XM_018053253.1 | XP_017908742.1 | 3 - 4 | 574 | 105 | ||
XM_018053254.1 | XP_017908743.1 | 4 - 5 | 617 | 105 | ||
XM_018053255.1 | XP_017908744.1 | 4 - 5 | 551 | 105 | ||
GenBank mRNA | KA346075.1 | - | 4 - 5 | 1312 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |