Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675231.3 | XP_005675288.3 | 1 - 2 | 602 | 96 | |
GenBank mRNA | JO362846.2 | - | 1 - 2 | 170 | 96 | |
JO415806.2 | - | 1 - 2 | 378 | 96 | ||
JO415807.2 | - | 1 - 2 | 378 | 96 | ||
JO430719.2 | - | 1 - 2 | 568 | 96 | ||
JO507354.2 | - | 3 - 4 | 661 | 96 | ||
JO583963.1 | - | 1 - 2 | 538 | 96 | ||
JO583964.1 | - | 1 - 2 | 538 | 96 | ||
JO641085.2 | - | 3 - 4 | 1226 | 96 | ||
JO641086.2 | - | 3 - 4 | 1226 | 96 | ||
JT705771.1 | - | 3 - 4 | 641 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |