Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053894.1 | XP_017909383.1 | 8 - 9 | 1474 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |