Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIRIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697676.3 | XP_005697733.2 | 1 - 2 | 99 | 67 | |
GenBank mRNA | JT670997.1 | - | 3 | 73 |
Set Membership: |
Amplicon length less than or equal to 70 Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |