Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059502.1 | XP_017914991.1 | 4 - 5 | 1724 | 88 | |
XM_018059509.1 | XP_017914998.1 | 4 - 5 | 1724 | 88 | ||
GenBank mRNA | JO637725.2 | - | 4 - 5 | 616 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |