Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059497.1 | XP_017914986.1 | 3 - 4 | 448 | 91 | |
XM_018059498.1 | XP_017914987.1 | 3 - 4 | 448 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons |