Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055031.1 | XP_017910520.1 | 3 - 4 | 533 | 64 | |
GenBank mRNA | JP350308.2 | - | 3 - 4 | 398 | 64 | |
JP350309.2 | - | 3 - 4 | 398 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |