Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692863.3 | XP_005692920.3 | 1 - 2 | 252 | 88 | |
XM_018062886.1 | XP_017918375.1 | 1 - 2 | 252 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |