Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285550.1 | NP_001272479.1 | 4 - 5 | 351 | 107 | |
GenBank mRNA | AB201469.1 | - | 4 - 5 | 351 | 107 | |
JO329459.2 | - | 4 - 5 | 430 | 107 | ||
JO348369.2 | - | 4 - 5 | 464 | 107 | ||
JO377483.2 | - | 4 - 5 | 460 | 107 | ||
JO420345.2 | - | 4 - 5 | 464 | 107 | ||
JO510611.2 | - | 4 - 5 | 450 | 107 | ||
JO538630.2 | - | 4 - 5 | 452 | 107 | ||
JO568887.2 | - | 4 - 5 | 444 | 107 | ||
JO580858.1 | - | 4 - 5 | 523 | 107 | ||
JO635060.2 | - | 1 - 2 | 444 | 107 | ||
JO635061.2 | - | 1 - 2 | 444 | 107 | ||
JP351188.2 | - | 4 - 5 | 456 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |