Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285657.1 | NP_001272586.1 | 4 - 5 | 303 | 60 | |
GenBank mRNA | EU244429.1 | - | 4 - 5 | 303 | 60 | |
EU244430.1 | - | 4 - 5 | 303 | 60 | ||
JO393062.2 | - | 3 - 4 | 486 | 60 | ||
KA343267.1 | - | 3 - 4 | 500 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |