Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285757.1 | NP_001272686.1 | 1 | 3115 | 60 | |
XM_013976677.2 | XP_013832131.1 | 3 | 3176 | 60 | ||
XM_013976679.2 | XP_013832133.1 | 2 | 3111 | 60 | ||
GenBank mRNA | GU936188.1 | - | 1 | 3027 | 60 | |
GU936189.1 | - | 1 | 3027 | 60 | ||
HM769860.1 | - | 1 | 3117 | 60 | ||
JT762513.1 | - | 1 | 1515 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |