Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285766.1 | NP_001272695.1 | 7 - 8 | 1302 | 101 | |
GenBank mRNA | AY510111.1 | - | 7 - 8 | 1302 | 101 | |
KA352949.1 | - | 1 - 2 | 542 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |