Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314243.1 | NP_001301172.1 | 2 - 3 | 518 | 72 | |
GenBank mRNA | JO438586.2 | - | 2 - 3 | 726 | 72 | |
JQ341163.1 | - | 2 - 3 | 518 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |