Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285744.1 | NP_001272673.1 | 4 - 5 | 568 | 90 | |
GenBank mRNA | GU985267.1 | - | 4 - 5 | 568 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |