Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286975.1 | NP_001273904.1 | 3 - 4 | 475 | 76 | |
XM_018063567.1 | XP_017919056.1 | 4 - 5 | 651 | 76 | ||
GenBank mRNA | HM038035.1 | - | 3 - 4 | 475 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |