Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314215.1 | NP_001301144.1 | 2 - 3 | 1535 | 69 | |
GenBank mRNA | JQ798185.1 | - | 2 - 3 | 1535 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |