Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285540.1 | NP_001272469.1 | 1 | 1022 | 98 | |
XM_018049146.1 | XP_017904635.1 | 2 | 1179 | 98 | ||
XM_018049147.1 | XP_017904636.1 | 2 | 1162 | 98 | ||
GenBank mRNA | HQ263211.1 | - | 1 | 1022 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |