Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285646.1 | NP_001272575.1 | 1 - 2 | 371 | 99 | |
XM_013967192.2 | XP_013822646.1 | 3 - 4 | 823 | 99 | ||
XM_013967193.2 | XP_013822647.1 | 3 - 4 | 584 | 99 | ||
XM_018053541.1 | XP_017909030.1 | 3 - 4 | 983 | 99 | ||
XM_018053542.1 | XP_017909031.1 | 3 - 4 | 857 | 99 | ||
GenBank mRNA | EF632080.1 | - | 1 - 2 | 371 | 99 | |
FJ194443.1 | - | 2 - 3 | 407 | 99 | ||
KA351327.1 | - | 3 - 4 | 990 | 99 | ||
KF492983.1 | - | 1 - 2 | 371 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |