Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286965.1 | NP_001273894.1 | 5 - 6 | 684 | 97 | |
GenBank mRNA | EU244428.1 | - | 5 - 6 | 418 | 97 | |
JO377265.2 | - | 5 - 6 | 684 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |