Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314281.1 | NP_001301210.1 | 3 - 4 | 340 | 96 | |
GenBank mRNA | JO603677.2 | - | 3 - 4 | 543 | 96 | |
JQ031152.1 | - | 3 - 4 | 340 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |