Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314350.1 | NP_001301279.1 | 2 - 3 | 216 | 72 | |
GenBank mRNA | GU446671.1 | - | 2 - 3 | 125 | 72 | |
JO647226.2 | - | 3 - 4 | 375 | 72 | ||
KM215210.1 | - | 2 - 3 | 216 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |