Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285539.1 | NP_001272468.1 | 2 - 3 | 277 | 91 | |
XM_018054687.1 | XP_017910176.1 | 2 - 3 | 453 | 91 | ||
XM_018054688.1 | XP_017910177.1 | 2 - 3 | 453 | 91 | ||
XM_018054689.1 | XP_017910178.1 | 2 - 3 | 453 | 91 | ||
GenBank mRNA | EF564255.1 | - | 2 - 3 | 91 | ||
JO517063.2 | - | 5 - 6 | 91 | |||
X58471.1 | - | 2 - 3 | 91 | |||
Z19569.1 | - | 2 - 3 | 277 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |