Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285613.1 | NP_001272542.1 | 2 - 3 | 276 | 70 | |
XM_013966068.2 | XP_013821522.2 | 3 - 4 | 842 | 70 | ||
XM_018051762.1 | XP_017907251.1 | 3 - 4 | 550 | 70 | ||
GenBank mRNA | EF151287.1 | - | 2 - 3 | 276 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |