Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287233.1 | NP_001274162.1 | 6 - 7 | 635 | 108 | |
XM_018047553.1 | XP_017903042.1 | 6 - 7 | 704 | 108 | ||
GenBank mRNA | JO581186.1 | - | 2042 | 108 | ||
JQ417404.1 | - | 6 - 7 | 635 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |