Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287576.1 | NP_001274505.1 | 1 - 2 | 156 | 81 | |
XM_005677829.3 | XP_005677886.2 | 2 - 3 | 188 | 81 | ||
GenBank mRNA | JQ937288.1 | - | 1 - 2 | 156 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |