Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314213.1 | NP_001301142.1 | 1 - 2 | 554 | 95 | |
GenBank mRNA | KA341451.1 | - | 1 - 2 | 1255 | 95 | |
KJ782301.1 | - | 1 - 2 | 554 | 95 | ||
KJ782304.1 | - | 1 - 2 | 554 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |