Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285638.1 | NP_001272567.1 | 2 - 3 | 278 | 95 | |
GenBank mRNA | AF003542.1 | - | 2 - 3 | 278 | 95 | |
AY603407.1 | - | 2 - 3 | 230 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |