Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285764.1 | NP_001272693.1 | 2 - 3 | 275 | 82 | |
XM_005694718.3 | XP_005694775.3 | 2 - 3 | 717 | 82 | ||
GenBank mRNA | HM138665.1 | - | 2 - 3 | 436 | 82 | |
HM138666.1 | - | 2 - 3 | 436 | 82 | ||
HM852511.1 | - | 2 - 3 | 275 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |