Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286091.1 | NP_001273020.1 | 2 - 3 | 183 | 88 | |
XM_018066621.1 | XP_017922110.1 | 3 - 4 | 376 | 88 | ||
GenBank mRNA | JO581437.1 | - | 3 - 4 | 1726 | 88 | |
JQ417405.1 | - | 2 - 3 | 183 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |