Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286092.1 | NP_001273021.1 | 1 - 2 | 375 | 83 | |
XM_018059989.1 | XP_017915478.1 | 1 - 2 | 426 | 83 | ||
GenBank mRNA | JO425752.1 | - | 1 - 2 | 157 | 83 | |
JO428925.2 | - | 1 - 2 | 97 | 83 | ||
JT717514.1 | - | 2 - 3 | 495 | 83 | ||
KF006348.1 | - | 1 - 2 | 375 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Capra hircus Probe spans exons |