Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286092.1 | NP_001273021.1 | 2 - 3 | 690 | 87 | |
XM_018059989.1 | XP_017915478.1 | 2 - 3 | 741 | 87 | ||
GenBank mRNA | JO425752.1 | - | 2 - 3 | 472 | 87 | |
JO428925.2 | - | 2 - 3 | 412 | 87 | ||
KF006348.1 | - | 2 - 3 | 690 | 87 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Capra hircus Probe spans exons |