Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286092.1 | NP_001273021.1 | 5 - 6 | 1932 | 106 | |
XM_018059989.1 | XP_017915478.1 | 5 - 6 | 2010 | 106 | ||
GenBank mRNA | JO425752.1 | - | 5 - 6 | 1714 | 106 | |
JO428925.2 | - | 5 - 6 | 1654 | 106 | ||
JT717514.1 | - | 6 - 7 | 2048 | 106 | ||
KF006348.1 | - | 5 - 6 | 1932 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Capra hircus Probe spans exons |