Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRINA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058698.1 | XP_017914187.1 | 6 - 7 | 1137 | 89 | |
GenBank mRNA | JO363369.2 | - | 6 - 7 | 1135 | 89 | |
JO431649.2 | - | 6 - 7 | 1122 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |