Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043098.1 | XP_017898587.1 | 3 - 4 | 622 | 72 | |
GenBank mRNA | JO580982.1 | - | 3 - 4 | 796 | 72 | |
KA327339.1 | - | 3 - 4 | 546 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |