Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674945.3 | XP_005675002.2 | 1 - 2 | 180 | 89 | |
GenBank mRNA | JO600865.2 | - | 4 - 5 | 1260 | 89 | |
JO600866.2 | - | 4 - 5 | 1374 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |