Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECTIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064224.1 | XP_017919713.1 | 2 - 3 | 667 | 114 | |
XM_018064281.1 | XP_017919770.1 | 2 - 3 | 667 | 114 | ||
GenBank mRNA | JO397501.2 | - | 4 - 5 | 1336 | 114 | |
JO444202.2 | - | 2 - 3 | 406 | 114 | ||
JO444979.2 | - | 2 - 3 | 406 | 114 | ||
JO573052.2 | - | 2 - 3 | 440 | 114 | ||
JT738668.1 | - | 1 - 2 | 270 | 114 | ||
KA340304.1 | - | 4 - 5 | 1027 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |