Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678735.3 | XP_005678792.2 | 2 - 3 | 554 | 109 | |
XM_013963012.2 | XP_013818466.2 | 2 - 3 | 625 | 109 | ||
XM_013963014.2 | XP_013818468.2 | 2 - 3 | 554 | 109 | ||
XM_018041131.1 | XP_017896620.1 | 2 - 3 | 540 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |