Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051295.1 | XP_017906784.1 | 2 - 3 | 1282 | 76 | |
XM_018051296.1 | XP_017906785.1 | 3 - 4 | 438 | 76 | ||
XM_018051297.1 | XP_017906786.1 | 2 - 3 | 1282 | 76 | ||
XM_018051298.1 | XP_017906787.1 | 2 - 3 | 1282 | 76 | ||
GenBank mRNA | JO617569.2 | - | 2 - 3 | 1341 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |