Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053153.1 | XP_017908642.1 | 3 - 4 | 421 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons |