Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053153.1 | XP_017908642.1 | 4 - 5 | 554 | 114 | |
GenBank mRNA | JT685674.1 | - | 4 - 5 | 776 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |