Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ONECUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697281.3 | XP_005697338.3 | 1 - 2 | 1418 | 54 | |
GenBank mRNA | JO409760.2 | - | 1 - 2 | 704 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |