Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038418.1 | XP_017893907.1 | 2 - 3 | 393 | 84 | |
GenBank mRNA | JO603249.2 | - | 2 - 3 | 379 | 84 | |
JO603250.2 | - | 2 - 3 | 813 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |