Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JT717645.1 | - | 2 - 3 | 70 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |