Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047106.1 | XP_017902595.1 | 4 - 5 | 1787 | 91 | |
GenBank mRNA | JO437490.2 | - | 4 - 5 | 619 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |