Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681272.3 | XP_005681329.2 | 4 - 5 | 3564 | 112 | |
XM_018049179.1 | XP_017904668.1 | 4 - 5 | 4156 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |