Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686085.3 | XP_005686142.2 | 3 - 4 | 1596 | 102 | |
XM_013967313.2 | XP_013822767.2 | 2 - 3 | 915 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons |